ENA333558649 Enamine

1-(2-chlorophenyl)-1-(4-ethylphenyl)methanamine

  • MDL number MFCD11148355
  • Cas Number : 1021126-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82897
Availability details for
Quantity
  • Availability
  • Add to Cart