ENA333558661 Enamine

(2-chlorophenyl)(2,4-dimethylphenyl)methanamine hydrochloride

  • MDL number MFCD19982456
  • Cas Number : 1333708-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268...270
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82912

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82912
Availability details for
Quantity
  • Availability
  • Add to Cart