ENA333558667 Enamine

2-azabicyclo[2.2.1]hept-5-en-3-one

  • Cas Number : 49805-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-58
Purity 95
Flash Point 110
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82926

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P301+P312,P304+P340,P312,P330,P403+P233,P405,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-82926
s_phrases_eu 26-36
pictograms GHS07
hstatements H302,H335
Availability details for
Quantity
  • Availability
  • Add to Cart