ENA333558671 Enamine

5-bromo-2-(2-hydroxy-2-methylpropoxy)benzaldehyde

  • MDL number MFCD12802053
  • Cas Number : 1184355-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82283
Availability details for
Quantity
  • Availability
  • Add to Cart