ENA333558674 Enamine

1-chloro-3-(isocyanatomethyl)benzene

  • MDL number MFCD06740483
  • Cas Number : 56620-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-82285
pictograms GHS07,GHS08
Availability details for
Quantity
  • Availability
  • Add to Cart