ENA333558677 Enamine

ethyl 5-[3-fluoro-2-(1H-imidazol-1-yl)phenyl]-1,2,4-oxadiazole-3-carboxylate

  • MDL number MFCD19982462
  • Cas Number : 1333915-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82950
Availability details for
Quantity
  • Availability
  • Add to Cart