ENA333558692 Enamine

N-(4-fluorophenyl)-1,3-dioxaindane-5-carboxamide

  • MDL number MFCD01269233
  • Cas Number : 349415-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-82976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82976
Availability details for
Quantity
  • Availability
  • Add to Cart