ENA333558693 Enamine

2-[(pyridin-3-yl)methoxy]benzoic acid

  • MDL number MFCD08310522
  • Cas Number : 120003-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82977
Availability details for
Quantity
  • Availability
  • Add to Cart