ENA333558708 Enamine

N-methyl-1-(pyridazin-3-yl)piperidin-4-amine

  • MDL number MFCD16767465
  • Cas Number : 1249245-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83004
Availability details for
Quantity
  • Availability
  • Add to Cart