ENA333558719 Enamine

[1-(thiophen-2-yl)cyclohexyl]methanamine

  • MDL number MFCD09693874
  • Cas Number : 767627-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83028

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83028
Availability details for
Quantity
  • Availability
  • Add to Cart