ENA333558720 Enamine

3-(butylsulfanyl)propanoic acid

  • MDL number MFCD01571819
  • Cas Number : 22002-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83029

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83029
Availability details for
Quantity
  • Availability
  • Add to Cart