ENA333558733 Enamine

N-[2-(pyrrolidin-1-yl)ethyl]aniline

  • MDL number MFCD09035368
  • Cas Number : 36716-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83053

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83053
Availability details for
Quantity
  • Availability
  • Add to Cart