ENA333558734 Enamine

1-(5-chloro-2-methoxyphenyl)-3-methylbutan-1-amine

  • MDL number MFCD14530571
  • Cas Number : 1215919-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82288
Availability details for
Quantity
  • Availability
  • Add to Cart