ENA333558740 Enamine

[(furan-2-yl)methyl][(5-methyl-1-benzofuran-2-yl)methyl]amine

  • MDL number MFCD16284419
  • Cas Number : 1272207-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83086
Availability details for
Quantity
  • Availability
  • Add to Cart