ENA333558753 Enamine

N-(2,3-dihydro-1-benzofuran-5-yl)piperidine-2-carboxamide hydrochloride

  • MDL number MFCD19982480
  • Cas Number : 1333644-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83105
Availability details for
Quantity
  • Availability
  • Add to Cart