ENA333558757 Enamine

4-{3-[(2-methoxyphenyl)methyl]-1,2,4-oxadiazol-5-yl}piperidine

  • MDL number MFCD19982482
  • Cas Number : 1291984-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83110
Availability details for
Quantity
  • Availability
  • Add to Cart