ENA333558769 Enamine

2-(1H-pyrazol-1-yl)pyridine-4-carboxylic acid

  • MDL number MFCD11128544
  • Cas Number : 1152940-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83140
Availability details for
Quantity
  • Availability
  • Add to Cart