ENA333558770 Enamine

2-(2-amino-1,3-thiazol-4-yl)phenol

  • MDL number MFCD00665956
  • Cas Number : 60135-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82298

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82298
Availability details for
Quantity
  • Availability
  • Add to Cart