ENA333558775 Enamine

ethyl 2-hydroxy-3-phenylpropanoate

  • MDL number MFCD00128177
  • Cas Number : 15399-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82299
Availability details for
Quantity
  • Availability
  • Add to Cart