ENA333558781 Enamine

2-amino-1-(8-chloro-2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-ol

  • MDL number MFCD11182087
  • Cas Number : 1094248-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83156
Availability details for
Quantity
  • Availability
  • Add to Cart