ENA333558783 Enamine

N-(3-aminophenyl)propane-1-sulfonamide

  • MDL number MFCD09048435
  • Cas Number : 926199-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82238
Availability details for
Quantity
  • Availability
  • Add to Cart