ENA333558797 Enamine

methyl 3-(3-bromophenyl)-2-methyl-3-oxopropanoate

  • MDL number MFCD16077019
  • Cas Number : 1247559-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83212

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83212
Availability details for
Quantity
  • Availability
  • Add to Cart