ENA333558813 Enamine

(3S)-3-carbamoyl-3-acetamidopropanoic acid

  • MDL number MFCD00237475
  • Cas Number : 60803-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82453

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82453
Availability details for
Quantity
  • Availability
  • Add to Cart