ENA333558815 Enamine

5-(methylsulfanyl)pentanoic acid

  • MDL number MFCD12823554
  • Cas Number : 69082-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83242
Availability details for
Quantity
  • Availability
  • Add to Cart