ENA333558816 Enamine

N,2-dimethylpyrimidin-4-amine

  • MDL number MFCD14704991
  • Cas Number : 33643-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-81
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-83243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83243
Availability details for
Quantity
  • Availability
  • Add to Cart