ENA333558819 Enamine

1-cyclohexyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD16666483
  • Cas Number : 1333542-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83259
Availability details for
Quantity
  • Availability
  • Add to Cart