ENA333558826 Enamine

2-(pyridin-2-yloxy)benzoic acid

  • MDL number MFCD13187322
  • Cas Number : 857540-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83266

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83266
Availability details for
Quantity
  • Availability
  • Add to Cart