ENA333558827 Enamine

1-(3-methylbutyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD16666454
  • Cas Number : 1267129-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187...189
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83267

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83267
Availability details for
Quantity
  • Availability
  • Add to Cart