ENA333558834 Enamine

4,4-dimethylpentan-1-amine hydrochloride

  • MDL number MFCD19982499
  • Cas Number : 1266694-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83278
Availability details for
Quantity
  • Availability
  • Add to Cart