ENA333558836 Enamine

2-(pyridin-2-ylformamido)acetic acid

  • MDL number MFCD08444886
  • Cas Number : 5616-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83280

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83280
Availability details for
Quantity
  • Availability
  • Add to Cart