ENA333558839 Enamine

2-(prop-2-en-1-yloxy)-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD17524447
  • Cas Number : 1333869-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83284
Availability details for
Quantity
  • Availability
  • Add to Cart