ENA333558857 Enamine

2-{[(tert-butoxy)carbonyl]amino}-2-(3-chlorophenyl)acetic acid

  • MDL number MFCD03426365
  • Cas Number : 669713-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83323
Availability details for
Quantity
  • Availability
  • Add to Cart