ENA333558861 Enamine

2-chloro-4-[(2,2-dimethylpropyl)amino]benzamide

  • MDL number MFCD14625244
  • Cas Number : 1250116-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-83336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83336
Availability details for
Quantity
  • Availability
  • Add to Cart