ENA333558862 Enamine

5-fluoro-2,3-dihydro-1-benzofuran-2-carboxylic acid

  • MDL number MFCD11186818
  • Cas Number : 89197-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83337
Availability details for
Quantity
  • Availability
  • Add to Cart