ENA333558865 Enamine

2-chloro-4-(4-methyl-1H-pyrazol-1-yl)pyridine

  • MDL number MFCD18885929
  • Cas Number : 1289021-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83340
Availability details for
Quantity
  • Availability
  • Add to Cart