ENA333558866 Enamine

1-bromo-3-(2,2,2-trifluoroethoxy)propane

  • MDL number MFCD11102050
  • Cas Number : 1152515-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-83344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83344
Availability details for
Quantity
  • Availability
  • Add to Cart