ENA333558869 Enamine

[2-(2,2,2-trifluoroethoxy)cyclohexyl]methanamine

  • MDL number MFCD12190142
  • Cas Number : 1155125-61-3
  • Molecular Weight : 211.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(2,2,2-trifluoroethoxy)cyclohexyl]methanamine
Inchi Code 1S/C9H16F3NO/c10-9(11,12)6-14-8-4-2-1-3-7(8)5-13/h7-8H,1-6,13H2
Inchi Key NGVIRIIQGBINNV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-83350

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-83350
pictograms GHS05,GHS07
hstatements H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart