ENA333558882 Enamine

3-amino-2-methylbutan-1-ol

  • MDL number MFCD18430999
  • Cas Number : 78401-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83414
Availability details for
Quantity
  • Availability
  • Add to Cart