ENA333558883 Enamine

5-methylhexan-1-amine

  • MDL number MFCD09923499
  • Cas Number : 4746-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83415
Availability details for
Quantity
  • Availability
  • Add to Cart