ENA333558892 Enamine

6-[(2-chlorophenyl)carbonyl]-2,3-dihydro-1,4-benzodioxine

  • MDL number MFCD00604246
  • Cas Number : 164526-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82466

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82466
Availability details for
Quantity
  • Availability
  • Add to Cart