ENA333558894 Enamine

2-(3-methylphenyl)propanoic acid

  • MDL number MFCD11036888
  • Cas Number : 73721-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-63
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82467
Availability details for
Quantity
  • Availability
  • Add to Cart