ENA333558903 Enamine

5-(piperidine-1-sulfonyl)pyridin-2-amine

  • MDL number MFCD06307747
  • Cas Number : 627839-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-83440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83440
Availability details for
Quantity
  • Availability
  • Add to Cart