ENA333558908 Enamine

3-phenoxybutan-2-amine

  • MDL number MFCD14626598
  • Cas Number : 6437-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-83445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83445
Availability details for
Quantity
  • Availability
  • Add to Cart