ENA333558916 Enamine

1-methyl-4-nitro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD00464004
  • Cas Number : 4598-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83464

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-83464
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart