ENA333558917 Enamine

4-iodo-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD00465278
  • Cas Number : 6647-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83465
Availability details for
Quantity
  • Availability
  • Add to Cart