ENA333558920 Enamine

4-bromo-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD00463988
  • Cas Number : 84547-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83468

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83468
Availability details for
Quantity
  • Availability
  • Add to Cart