ENA333558921 Enamine

4-chloro-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD01821187
  • Cas Number : 84547-85-3
  • Molecular Weight : 160.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-1-methyl-1H-pyrazole-3-carboxylic acid
Inchi Code 1S/C5H5ClN2O2/c1-8-2-3(6)4(7-8)5(9)10/h2H,1H3,(H,9,10)
Inchi Key BZFXEYAWDBWTSY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-83469

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-83469
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart