ENA333558923 Enamine

1-ethyl-4-nitro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD02090836
  • Cas Number : 401906-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83471
Availability details for
Quantity
  • Availability
  • Add to Cart