ENA333558930 Enamine

4-[(4-bromo-1H-pyrazol-1-yl)methyl]benzoic acid

  • MDL number MFCD01893318
  • Cas Number : 387344-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83478
Availability details for
Quantity
  • Availability
  • Add to Cart