ENA333558932 Enamine

5-(4-chloro-3-methylphenoxymethyl)furan-2-carboxylic acid

  • MDL number MFCD02090846
  • Cas Number : 406470-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83480

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83480
Availability details for
Quantity
  • Availability
  • Add to Cart